Learn More
Tocris Bioscience™ (±)-ML 209
RORγt inverse agonist; suppresses Th17 cell differentiation
Brand:  Tocris Bioscience™ 5987/50
 View more versions of this product
Product Code. 18736658
  • 733.00 € / 50mg
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
(±)-ML 209 is a RORγt inverse agonist (IC50 = 460 nM). Suppresses Th17 cell differentiation and IL17A expression in vitro. Does not suppress transcription of RORα. Minimal activity displayed on ERα, LXRα and thyroid hormone receptors (IC50 values >4.4 μM).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
White
1334526-14-5
441.52g/mol
Store at 4°C
50 mg
Inhibitor
C25H31NO6
Soluble to 100mM in DMSO
Agonist against RORγt
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Special Offers
Special Offers
For Laboratory Research Use Only