missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 365
Potent and selective K2P3.1 (TASK-1) channel blocker
248.00 € - 978.00 €
Specifications
Purity | >99% |
---|---|
Product Type | ML 365 |
Molecular Weight (g/mol) | 360.41 |
Inhibitors | ML 365 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18736158
|
Tocris Bioscience™
5337/10 |
10 mg |
248.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18754729
|
Tocris Bioscience™
5337/50 |
50 mg |
978.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent and selective TASK-1 (K2P3.1/KCNK3) channel blocker (IC50 values are 4 and 390 nM at TASK-1 and TASK-3, respectively). Displays little or no inhibition at Kir2.1, voltage-gated potassium channels, KCNQ2 and KV11.1 (hERG).Specifications
>99% | |
ML 365 | |
360.41 | |
ML 365 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title