Learn More
ML171, MedChemExpress
MedChemExpress ML171 (2-Acetylphenothiazine;2-APT) is a potent and selective NADPH oxidase 1 (Nox1) inhibitor that blocks Nox1-dependent ROS generation, with an IC50 of 0.25 μM in HEK293-Nox1 confirmatory assay.
Brand:  Medchem Express HY-12805
Encompass_Preferred
 View more versions of this product
Product Code. 16465986
  • 41.85 € / 100mg
Explore more special offers
Description
Description
ML171 (2-Acetylphenothiazine
Specifications
Specifications
ML171
Orange
C14H11NOS
2-Acetylphenothiazine 2-APT
CC(C1=CC=C2SC3=CC=CC=C3NC2=C1)=O
241.31
Research
COVID-19-immunoregulation
6631-94-3
Research
100 mg
DMSO : ≥ 64 mg/mL (265.22 mM)
241.31
99.9%
Solid