missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | MLH1 |
---|---|
Concentration | 0.4 mg/mL |
Applications | Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16858953
|
Proteintech
11697-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16848953
|
Proteintech
11697-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is specific to MLH1.
MLH1 is a DNA mismatch repair protein. The repair of mismatch DNA is essential to maintaining the integrity of genetic information over time. An alteration of microsatellite repeats is the result of slippage owing to strand misalignment during DNA replication and is referred to as microsatellite instability (MSI). These defects in DNA repair pathways have been related to human carcinogenesis. The importance of mismatch repair genes became apparent with the identification of the genetic basis for hereditary nonpolyposis colon cancer (HNPC). MSHS2 is involved in the initial cognition of mismatch nucleotides during the replication mismatch repair process. It is thought that after MSH2 binds to a mismatched DNA duplex it is joined by a heterodimer of MLH1 and PMSH, which together help facilitate the later steps in mismatch repair.Specifications
MLH1 | |
Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
P40692, P97679, Q9JK91 | |
17350, 4292, 81685 | |
MLH1 Fusion Protein Ag2319 | |
Primary | |
-20°C | |
MLH1 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
COCA2, FCC2, hMLH1, HNPCC, HNPCC2, MLH1, MutL protein homolog 1 | |
Mlh1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title