Learn More
Tocris Bioscience™ MM 102
WDR5/MLL interaction inhibitor
Brand:  Tocris Bioscience™ 5307/10
226.34 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18793968
  • 296.00 € / 10mg
Explore more special offers
Description
Description
MM 102 is a potent WDR5/MLL interaction inhibitor (IC50 = 2.4 nM). Inhibits MLL1 methyltransferase activity and MLL-1-induced HoxA9 and Meis-1 gene expression in leukemia cells expressing the MLL1-AF9 fusion gene. Also inhibits cell growth and induces apoptosis in leukemia cells harbouring MLL1 fusion proteins.
Specifications
Specifications
>98%
MM 102
MM 102
1883545-52-5
783.83