Learn More
MMAE-d8, MedChemExpress
MedChemExpress MMAE-d8 (Monomethyl auristatin E-d8) is a deuterated labeled MMAE, a potent mitotic inhibitor and a tubulin inhibitor.
Brand:  Medchem Express HY-15162A
Encompass_Preferred
 View more versions of this product
Product Code. 16491479
  • 1841.40 € / 1mg
Explore more special offers
Description
Description
MMAE-d8 (Monomethyl auristatin E-d8) is a deuterated labeled MMAE, a potent mitotic inhibitor and a tubulin inhibitor.
Specifications
Specifications
MMAE-d8
1 mg
Research
Monomethyl auristatin E-d8 Deuterated labeled MMAE
CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C)[C@@H](O)C2=CC=CC=C2)=O)OC)=O)OC)N(C([C@@](C(C([2H])([2H])[2H])(C([2H])([2H])[2H])[2H])([2H])NC([C@H](C(C)C)NC)=O)=O)C
726.03
Research
Solid
2070009-72-0
White
C39H59D8N5O7
DMSO : 100 mg/mL (137.74 mM; Need ultrasonic)
726.03
98.43%
Cancer-programmed cell death