missing translation for 'onlineSavingsMsg'
Learn More

MMAF-d8 hydrochloride, MedChemExpress

MedChemExpress D8-MMAF hydrochloride is a deuterated form of MMAF hydrochloride, which is a microtubule disrupting agent.

Encompass_Preferred

1422.90 € - 6066.90 €

Specifications

Chemical Name or Material MMAF-d8 hydrochloride
Color Off-White
Purity Grade Notes Research
Molecular Formula C39H58D8ClN5O8
Solubility Information DMSO : 100 mg/mL (128.79 mM; Need ultrasonic)
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16471529
Encompass_Preferred
View Documents
Medchem Express
HY-15579AS
1 mg
1422.90 €
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16449207
Encompass_Preferred
View Documents
Medchem Express
HY-15579AS
5 mg
3207.60 €
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16436648
Encompass_Preferred
View Documents
Medchem Express
HY-15579AS
10 mg
6066.90 €
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

D8-MMAF hydrochloride is a deuterated form of MMAF hydrochloride, which is a microtubule disrupting agent.
Specifications

Specifications

MMAF-d8 hydrochloride
Research
DMSO : 100 mg/mL (128.79 mM; Need ultrasonic)
Solid
776.47
98.97%
Off-White
C39H58D8ClN5O8
Cancer-programmed cell death
CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C(O)=O)CC2=CC=CC=C2)=O)OC)=O)OC)N(C([C@@](C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])([2H])NC([C@H](C(C)C)NC)=O)=O)C.Cl
776.47
Research
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣The compound is unstable in solutions, freshly prepared is recommended.

Recommended Storage : 4°C, sealed storage, away from moisture∣The compound is unstable in solutions, freshly prepared is recommended.

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
MMAF-d8 hydrochloride, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.