Learn More
Macherey-Nagel™ MN 615 Grade Round Format Cellulose Qualitative Filter Paper
Use for a wide range of general laboratory filtrations. MN 615 Grade Round Format Cellulose Qualitative Filter Paper provides medium fast filtration.
Brand:  Macherey-Nagel™ 431005
 View more versions of this product
Product Code. 10597511
  • 6.30 € / Pack of 100
Explore more special offers
Description
Description
  • For general purpose filtrations
  • Qualitative filter papers are manufactured from the same raw materials as the ashless grades and are particularly suited for general laboratory filtrations
  • The average ash content is about 0.1%
  • The amount of a-cellulose is about 95%.
  • Medium fast filtration
Specifications
Specifications
Disc
Medium fast
MN 615
0.16 mm
55 mm
Cellulose
Qualitative analysis
100 Pack
Filter Paper