Learn More
Macherey-Nagel™ MN 640 W Filter Paper
Brand:  Macherey-Nagel™ 202901
Product Code. 11308460
  • 63.20 € / Pack of 100
Explore more special offers
Specifications
85 g/m2
100Pack
Filter Paper
For quantitative analysis and filtration with low blank values
0.2 mm