Learn More
IKA™ R 2850 Mobile Floor Stand
Mobile floor stand, with H-shape base which prevents against tipping.
Brand: IKA™ 0020002900
This item is not returnable.
View return policy
Description
For overhead stirrers and dispersers with a diameter of extension arm of 16 mm.Specifications
875 mm | |
16 mm | |
For Overhead Stirrers and Dispersers With a Diameter of Extension Arm of 16mm | |
1 Ea. | |
38.6 in. |
0.6 in. | |
For Overhead Stirrers and Dispersers With a Diameter of Extension Arm of 16mm | |
1921 mm | |
Mobile Floor Stand | |
980 mm |