Learn More
Miele Professional™ Mobile Injector
Mobile unit for optimum loading of pipettes
Brand:  Miele Professional™ 6727920
Product Code. 10683191
  • 2360.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Equipped with 38 pipettes in 3 rows
  • 1st Row: 10 pipettes x 100 ml (up to 550 mm), bar distance 20 mm
  • 2nd Row: 14 pipettes x 25 ml , bar distance 26 mm
  • 3rd Row: 14 pipettes x 10 ml, bar distance 26 mm
  • Bracket for ML/2 for automatic mobile unit recognition
Specifications
Specifications
Mobile Injector Unit with Drying Connection
E 405/1
Pipettes
1 Each