missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Mobile injector

Module E 947/2 for cleaning 88 centrifuge tubes, phials or test tubes, for example

Brand:  Miele Professional™ 7459430

Product Code. 15400565

  • 3290.00 € / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Equipped with 88 nozzles, 2.5 x 110 mm
Specifications

Specifications

Stainless Steel, Plastic
Stainless Steel, Black
Washer-disinfectors, laboratory
170 mm
Insert/Module for Test Tubes
88 Test tubes
352 mm
558 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.