missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Miele Professional™ Mobile Injector, Module A 300
Module A 300 for the optimum loading of laboratory glassware.
Brand: Miele Professional™ 11116900
333.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Suitable for laboratory glassware such as Erlenmeyer flasks
- Equipped with 8 injector nozzles E 352 (6 x 220 mm)
- Equipped with 8 locking mechanisms for nozzles E 354
- Jet spacing 120 mm across the width, 123 mm across the depth

Specifications
Injector Module | |
Laboratory Washer Baskets A 100 and A 150 | |
Two rows of four injectors | |
181 x 220 x 479 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Miele Professional™ Mobile Injector, Module A 300 > For Use With: Laboratory washer baskets A 100 and A 150
Spot an opportunity for improvement?Share a Content Correction