Learn More
DuPont™ Tyvek™ 500 Xpert Hooded Coverall Green
GREENER_CHOICE
The Tyvek™ 500 Xpert Hooded Coverall is composed of flash-spun HDPE, providing an ideal balance of protection, durability and comfort. The 3-piece hood provides optimal fit to head and face when turning while ample crotch area offers freedom of movement.
955.00 €
Specifications
Product Type Hooded Coveralls
Material HDPE
Color Green
Waist Style Elastic
Ankle Style Elastic
View More Specs
Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
13436646
GREENER_CHOICE
View accessory products View Documents Promotion Details
DuPont™
D14936675
X-Large
955.00 €
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 09-10-2024
to see stock.
Add to basket
13416646
GREENER_CHOICE
View accessory products View Documents Promotion Details
DuPont™
D14936650
Medium
955.00 €
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 09-10-2024
to see stock.
Add to basket
13456646
GREENER_CHOICE
View accessory products View Documents Promotion Details
DuPont™
D14936681
2X-Large
955.00 €
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
13446646
GREENER_CHOICE
View accessory products View Documents Promotion Details
DuPont™
D14936664
Large
955.00 €
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 09-10-2024
to see stock.
Add to basket
Description
Description
  • Coverall with 3-piece hood for optimal fit to head and face when turning.
  • Excellent barrier against fine particles and fibers down to 1 micron in size.
  • Ergonomic-protective design.
  • Stitched external seams.
  • Elasticated face, wrists and ankles as well as glued-in waist elastic.
  • Large, easy-to-grasp zipper puller.
  • Robust yet lightweight (<180 g per garment).
  • Ample crotch area for freedom of movement.
  • Ultra-low-linting and antistatically treated.
Specifications
Specifications
Hooded Coveralls
Green
Elastic
Hooded
Zipper
Category III, Type 5 and 6
(EU) 2016/425, EN 14126, EN 1073-2
HDPE
Elastic
Elastic
3-panel
Stitched
Abrasion