Learn More
DuPont™ Tyve 600 Plus Green Hooded Coverall
Providing an ideal balance of protection, durability and comfort. DuPont™ Tyvek™ 600 Plus Green Hooded Coveralls are composed of flash spun high density polyethylene.
1130.00 €
Specifications
Product Type Hooded Coveralls
Material Tyvek
Color Green
Waist Style Elastic
Ankle Style Elastic
View More Specs
Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
11853510
View accessory products View Documents
DuPont™
D13495709
Large
1130.00 €
Pack of 100
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
12653720
View accessory products View Documents
DuPont™
D13495686
2X-Large
1130.00 €
Pack of 100
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
12663720
View accessory products View Documents
DuPont™
D13495715
Medium
1130.00 €
Pack of 100
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
12673720
View accessory products View Documents
DuPont™
D13495738
X-Large
1130.00 €
Pack of 100
Estimated Shipment: 01-05-2025
to see stock.
Add to basket
Description
Description
  • Self-adhesive chin strap for tight seal of suit to face mask
  • Stitched and over-taped seams for protection and strength
  • Tyvek zipper and zipper flap for enhanced protection
  • Elasticated face, wrists and ankles as well as a glued-in elastic waist
  • Elasticated thumb loops prevent sleeves from riding up
  • Permeable to both air and water vapour, yet repels water-based liquids and aerosols
  • Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
  • Applications include maintenance and dismantling jobs in the nuclear industry, pharmaceutical manufacturing or research, biosecurity laboratories as well as in medical applications and when exposed to biological hazards
Specifications
Specifications
Hooded Coveralls
Green
Elastic
Hooded
Zipper with Storm Flap
Category III, Type 4-B, 5-B and 6-B
(EU) 2016/425, EN 14126
<250 g
Tyvek
Elastic
Elastic
Elastic
Stitched, Taped
Self-adhesive chin flap
Applications for Tyvek™ Classic Plus garments include maintenance and dismantling jobs in the nuclear industry, pharmaceutical manufacturing or in research and biosecurity laboratories, as well as in medical applications and when exposed to biological hazards
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
DuPont™ Tyve 600 Plus Green Hooded Coverall