missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Modified Harris Hematoxylin
Use Epredia™ Modified Harris Hematoxylin, a regressive nuclear stain, for both histology and cytology.
81.50 € - 85.10 €
Description
- Formulation contains no mercury
- More stable than conventional Harris Hematoxylin
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Carcinogenicity Category 1A
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H350-May cause cancer.
Precautionary Statement
- P362-Take off contaminated clothing.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : hematoxylin, sodiumiodate, ethyl alcohol, methyl alcohol, diazolidinyl urea, acetic acid, aluminium
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title