Learn More
Corning™ Modified Impeller Assembly
Stainless steel impeller shaft with modified impeller blade for use with probes to create a small bioreactor.
Brand: Corning™ 402648
This item is not returnable.
View return policy
Description
Stainless steel impeller shaft with modified impeller blade for use with probes to create a small bioreactor.
Specifications
Impeller shaft with dual bearing | |
Stainless Steel | |
Magnetically-Driven 3L Proculture Spinner Flask | |
Non-sterile |
Product Content Correction
Product Title
Corning™ Modified Impeller Assembly > For use with Magnetically-Driven 3L Proculture Spinner Flask
Spot an opportunity for improvement?Share a Content Correction