Learn More
Miele Professional™ Mobile Injector, Module A 306
Module A 306 for the optimum loading of large volume laboratory glassware
Brand:  Miele Professional™ 9862580
Product Code. 15455021
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Suitable for 4 1-2 litre measuring cylinders, high shape
  • Suitable for 2 1-2 litre measuring cyl., low shape + 2 high shape
  • Contact surfaces plastic coated