Learn More
Thermo Scientific™ Molecular BioProducts™ HLT Ergo-F Pipet Tips
GREENER_CHOICE
For use with Finnpipette Focus and Digital Pipetters
Brand:  Thermo Scientific™ 3812
Product Code. 12924077
  • 58.60 € / Pack of 960
Estimated Shipment: 15-12-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ergonomic tips reduce strains and injury
  • Secure and consistent seals
  • Less ejection force
  • Also available in low retention polymer
TRUSTED_SUSTAINABILITY
Specifications
Specifications
200 μL
Rack
Molecular BioProducts HLT Ergo-F
Non-sterile
Finnpipette™ Focus, Digital Pipette