Learn More
3M™ Molecular Detection Assay 2
Used with the 3M™ Molecular Detection System for the rapid and specific detection of Cronobacter species in enriched powdered infant formula, raw materials to manufacture powdered infant formula, infant cereals, and environmental samples.
Brand:  3M™ 7100104558
Encompass
Product Code. 17278540
  • POA / 1 set
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees.
  • Ready-to-use and pre-dispensed reagents provide convenience and minimize the chance of human error.
  • Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification.
  • The ability to run all assays simultaneously and the flexibility to test up to 96 samples in each run helps you operate more efficiently.
  • Real-time amplification and detection process identifies positive results in as early as 15 minutes and allows you to make critical decisions faster.
Specifications
Specifications
Cronobacter MDA2CRO96
Microbiology Media
96
Cronobacter species
Environmental Test
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
3M™ Molecular Detection Assay 2