Learn More
3M™ Molecular Detection Assay 2
Used with the 3M™ Molecular Detection System for the rapid and specific detection of shiga toxin-producing e.coli species in enriched food and environmental samples.
POA
Specifications
Description STEC Gene Screen
Product Type Microbiology Media
Test Type Food and Beverage, Environmental Test
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17218550
Encompass
View Documents
3M™
7100194012
96 Pack
POA
1 set
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
17228550
Encompass
View Documents
3M™
7100194040
48 Pack
POA
1 set
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees.
  • Ready-to-use and pre-dispensed reagents provide convenience and minimize the chance of human error.
  • Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification.
  • The ability to run all assays simultaneously and the flexibility to test up to 96 samples in each run helps you operate more efficiently.
  • Real-time amplification and detection process identifies positive results in as early as 15 minutes and allows you to make critical decisions faster.
Specifications
Specifications
STEC Gene Screen
Microbiology Media
Food and Beverage, Environmental Test