missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Molecular Detection Cap/Decap Tool
Molecular detection cap and decap tool with lysis and reagent
624.32 € - 769.97 €
Specifications
Product Type | Accessory |
---|---|
Test Type | Food and Beverage, Environmental Test |
Quantity | 1 |
Product Code | Brand | Form | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Price | Quantity & Availability | |||||
17233744
|
3M™
7100001305 |
Reagent MDSCDR |
769.97 €
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
17243744
|
3M™
7100001306 |
Lysis MDSCDL |
624.32 €
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
Description
- Easy to use custom 3M accessory.
- Increase technician productivity.
Specifications
Accessory | |
Food and Beverage, Environmental Test | |
1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title