missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Amplex™ Red and Amplex™ UltraRed Reagents

The Amplex Red and UltraRed reagents are fluorogenic substrates for peroxidase. These highly sensitive and stable probes react with H2O2 in the presence of HRP for solution-based assays and ELISAs. Amplex UltraRed is a brighter version of Amplex Red with an improved pH range and sensitivity.

Brand:  Invitrogen™ A12222

 View more versions of this product

Product Code. 11519056

  • 333.00 € / 5mg

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Store in freezer (-5°C to -30°C).
Hydrogen Peroxidase Assay
Red Hydrogen Peroxide
Solid
5 mg
HRP (Horseradish Peroxidase) Substrate
Cell Analysis
Fluorescence
Fluorometer, Microplate Reader
Amplex Red Reagent
Amplex™
Room Temperature
Chemical substrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2615853 Certificate TypeCertificate of Analysis Date07/06/2023 Catalog NumberA12222
Lot Number2491409 Certificate TypeCertificate of Analysis Date24/05/2022 Catalog NumberA12222
Lot Number2311891 Certificate TypeCertificate of Analysis Date30/06/2021 Catalog NumberA12222
Lot Number2256708 Certificate TypeCertificate of Analysis Date08/10/2020 Catalog NumberA12222
Lot Number2140274 Certificate TypeCertificate of Analysis Date01/11/2019 Catalog NumberA12222
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Amplex™ Red and Amplex™ UltraRed Reagents > Amplex Red Reagent, 5 mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.