missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Amplex™ Red Xanthine/Xanthine Oxidase Assay Kit

Suitable for cell viability, proliferation and Function, cellular imaging, and enzyme and protein activity assays

Brand:  Invitrogen™ A22182

Product Code. 10207622

  • 630.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Provides sensitive and simple fluorometric method for detecting as little as 200nM xanthine or hypoxanthine or as little as 0.1mU/mL of xanthine oxidase activity in a purified system with a 100μL assay volume

Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Enzyme and Protein Activity Assays, High-Content Screening (HCS), Nitro-Oxidative Stress, Oxidase and Peroxidase Activity, Proteins, Expression, Isolation and Analysis, Reactive Oxygen Species

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer -5°C to -30°C and protect from light.
Xanthine/Xanthine Oxidase Assay
Red Xanthine/Xanthine Oxidase Assay Kit
Tube(s), 96-well plate
400 assays
Fluorescence
Microplate Reader, Spectrophotometer, Fluorometer
Other Label(s) or Dye(s)
Amplex™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.