missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calcium Orange™, AM, cell permeant - Special Packaging
Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+
Brand: Invitrogen™ C3015
374.68 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
- These indicators have uses in many calcium signaling investigations, including measuring intracellular Ca2+, following Ca2+ influx and release, and multiphoton excitation imaging of Ca2+ in living tissues
- Cells may be loaded with the AM ester forms of these calcium indicators by adding the dissolved indicator directly to dishes containing cultured cells
- Fluorescence signal from these cells is generally measured using fluorescence microscopy, fluorescence microplate assays, or flow cytometry
- Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
- These probes are excited by visible light, and because energy required for excitation is low, potential for cellular photodamage is reduced
- Commonly used laser-based instruments (i.e., confocal laser scanning microscopes) are able to efficiently excite these indicators, and their emissions are in regions of the spectrum where cellular autofluorescence and scattering backgrounds are often less of a problem
- Label (Ex/Em): Calcium Orange (549/576nm)
- Fluorescence intensity increase upon binding Ca2+ : approximately 3 fold
Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling
Order Info
Shipping Condition: Room temperature
Specifications
Store in freezer -5°C to -30°C and protect from light. | |
Cell Viability and Proliferation | |
Calcium Indicator | |
CALCIUM ORANGE™ | |
Room Temperature |
Fluorescence | |
Fluorescence Microscope | |
Fluorescent Dye-Based | |
10 x 50 μg |
For Research Use Only. Not for use in diagnostic procedures.