missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellEvent™ Caspase-3/7 Green ReadyProbes™ Reagent

Product Code. 13599180
Click to view available options
Quantity:
1.75 mL kit
Unit Size:
1.75mL
This item is not returnable. View return policy

Product Code. 13599180

Brand: Invitrogen™ R37111

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

1 x 1.75mL dropper bottles

Fluorogenic, no-wash indicator for live- and fixed-cell applications

Activation of caspase-3 is an early indicator of apoptosis.

  • Allows rapid and sensitive detection of cells destined for cell death
  • Formulated as room temperature-stable solution and comes in convenient dropper bottle: tip and drip2 drops/mL to stain cells
  • Optimized caspase-3/7 substrate for apoptosis analysis
  • Simple and fast no-wash protocol helps preserve delicate apoptotic cells
  • Compatible with both live-cell fluorescence imaging and formaldehyde-based fixation methods
  • Ready-to-use liquid formulation: no need to dilute, weigh, or pipet
  • Four amino acid peptide (DEVD) conjugated to a nucleic acid-binding dye that is nonfluorescent when not bound to DNA: The Caspase-3/7 Green reagent is intrinsically nonfluorescent, as DEVD peptide inhibits binding of dye to DNA
  • Upon activation of caspase-3/7 in apoptotic cells, DEVD peptide is cleaved and free dye can bind DNA, generating bright green fluorescence
  • Apoptotic cells with activated caspase-3/7 will have bright green nuclei, while cells without activated caspase 3/7 will show minimal fluorescence
  • Fluorescent signal survives fixation and permeabilization, allowing end-point assays and immunocytochemistry-based methods
  • Optimization may be needed for some cell types, conditions, and applications: In such cases, simply add more or fewer drops until optimal staining intensity is obtained
  • If desired, combine with red or far-red nuclear stain for total cell count
  • Excited with maximum at 502nm and has emission maximum at 530nm
  • It is detected through standard GFP and FITC filters

Apoptosis, Apoptosis Related Factors, Caspase Activation, Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, High-Content Screening (HCS)

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1.75 mL kit
Format Dropper Bottle
Product Type Caspase 3/7 Reagent
Content And Storage 1 × 1.75 mL dropper bottles

Store at ≤ 25°C.
Compatible Cells Mammalian Cells
Excitation/Emission 502/530
Color Green
Product Line CellEvent, ReadyProbes
Detection Method Fluorescence
For Use With (Application) Cell Imaging
For Use With (Equipment) Confocal Microscope, Floid™ Cell Imaging System, Fluorescence Microscope, Flow Cytometer, Floid™ Cell Imaging System
Label Type Other Label(s) or Dye(s)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.