missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellROX™ Green Reagent, for oxidative stress detection

Novel fluorogenic probe for measuring oxidative stress in live cells

Brand:  Invitrogen™ C10444

480.32 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.


600.40 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


Product Code. 12648335

  • 640.00 € / 250µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Five single-use vials with enough total reagent for five 96-well plates or 100 coverslips
This item is not returnable. View return policy

Description

Description

Cell-permeant dye is weakly fluorescent while in reduced state and exhibits bright green photostable fluorescence upon oxidation by reactive oxygen species (ROS) and subsequent binding to DNA, with absorption/emission maxima of ∽485/520 nm. This reagent can be formaldehyde-fixed and its signal survives detergent treatment, which makes it multiplexable with other compatible dyes and antibodies.CellROXGreen Reagent is compatible with various platforms, such as traditional fluorescence microscopy, high content screening (HCS), flow cytometery, and microplate-based fluorimetry or high throughput screening (HTS) as well as benchtop instruments such as FLoid, Tali, and Attune instruments.

  • Compatible with other live cell dyes, antibodies, and in cells expressing RFP
  • Provided as stable, ready-to-use DMSO solution with simple no-wash protocol compatible with standard workflows in fluorescence microscopy
  • Oxidative stress results from an imbalance between production of ROS and ability of cells to scavenge them
  • ROS play important role in the progression of several diseases, including inflammation, atherosclerosis, aging, and age-related degenerative disorders
  • Form: Frozen
  • Excitation Class: Visible

Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Flow Cytometry, Flow Cytometry Viability and Cytotoxicity Assays, Flow Cytometry of Cellular Processes, High-Content Screening (HCS), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Nitro-Oxidative Stress, Reactive Oxygen Species

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Green
5 x 50 μL
Imaging, HCS, Cytometer
Live Cell Imaging
Oxidative stress
Frozen
Store at ≤–20°C. Protect from light and desiccate.

2.5 mM stabilized solution in DMSO
485/520 nm
CellROX™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3080143 Certificate TypeCertificate of Analysis Date23/12/2024 Catalog NumberC10444_LITL18 PROMO,C10444
Lot Number3080077 Certificate TypeCertificate of Analysis Date06/11/2024 Catalog NumberC10444_LITL18 PROMO,C10444
Lot Number3017346 Certificate TypeCertificate of Analysis Date28/09/2024 Catalog NumberC10444_LITL18 PROMO,C10444
Lot Number2942301 Certificate TypeCertificate of Analysis Date31/07/2024 Catalog NumberC10444_LITL18 PROMO,C10444
Lot Number2980637 Certificate TypeCertificate of Analysis Date31/07/2024 Catalog NumberC10444_LITL18 PROMO,C10444
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ CellROX™ Green Reagent, for oxidative stress detection > 5 x 50uL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.