missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Click-iT™ EdU Alexa Fluor™ 647 HCS Assay
GREENER_CHOICE

Uses a highly specific click reaction

822.00 € - 3170.00 €

Specifications

Color Far-Red
Detection Method Fluorescence
For Use With (Application) High-content Screening (HCS)
For Use With (Equipment) High Content Analysis Instrument
Product Type HCS Assay
View More Specs

Includes: The kit contains sufficient material for two 96-well plates.

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10308974
GREENER_CHOICE
View Documents Promotion Details
Invitrogen™
C10357
10 x 96 well plates
3170.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10053294
GREENER_CHOICE
View Documents Promotion Details
Invitrogen™
C10356
2 x 96 well plates
822.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Click-iT EdU Alexa Fluor 647 HCS Assay is a superior alternative to traditional proliferation assays that is optimized for high-content imaging applications. In this assay the modified thymidine analogue EdU is efficiently incorporated into newly synthesized DNA and fluorescently labeled with a bright, photostable Alexa Fluor dye in a fast, highly specific click reaction. This fluorescent labeling of proliferating cells is accurate and compatible with antibody methods due to the mild click protocol. The most accurate proliferation-detection methods are based on the incorporation and measurement of nucleoside analogues in newly synthesized DNA, with bromodeoxyuridine (BrdU) a commonly used analogue. BrdU-labeled DNA is quantitated using anti-BrdU antibodies following DNA denaturation by harsh methods (HCl, heat, or enzymes) to expose the BrdU molecules. This step is time consuming and difficult to perform consistently. The harsh treatment can also adversely effect sample integrity and quality, which makes co-staining with other antibodies challenging.

  • Simple: works the first time, every time, in less time
  • Efficient: no denaturation steps or harsh treatment required
  • Content-rich results: better preservation of cell morphology, antigen structure, and DNA integrity
  • Consistent: not dependent on variable antibody lots for detection
  • Less hazardous

Superior Proliferation Methodology
The Click-iT EdU Alexa Fluor 647 HCS Assay provides a superior alternative to BrdU assays for measuring cell proliferation. EdU (5-ethynyl-2'-deoxyuridine) is a nucleoside analog of thymidine and is incorporated into DNA during active DNA synthesis. With Click-iT EdU, mild fixation and detergent permeabilization is sufficient for the small molecule-based Click-iT EdU detection reagent to gain access to the DNA. As a consequence, the Click-iT EdU HCS assay is not only easy to use, but more accurate and compatible with cell cycle analysis and other intracellular or extracellular targets for truly content-rich results.

The Click-iT assay can be used on cells in culture or in vivo following administration of EdU by feeding or injection methods. The Click-iT assay can be used with BrdU in dual pulse experiments by using the ant-BrdU (clone MoBu-1) antibody, which does not cross react with EdU. The Click-iTtechnology is compatible with immunohistochemical, immunocytochemical, and fluorescent dyes that are fixation tolerant or designed for fixed-cell labeling.

Cell Analysis, Cell Cycle, Cell Proliferation, Cell Viability, Proliferation and Function, Cellular Imaging, DNA Replication and Repair, High-Content Screening (HCS)

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Far-Red
High-content Screening (HCS)
HCS Assay
near IR
Less hazardous
Room Temperature
Fluorescence
High Content Analysis Instrument
Alexa Fluor™ 647
96-well plate
Alexa Fluor™, Click-iT™, Molecular Probes™
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.