missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Click-IT™ Tetramethylrhodamine (TAMRA) Protein Analysis Detection Kit
GREENER_CHOICE

Supports differential analysis of glycoprotein subclasses, total proteins and total glycoproteins in the same gel

Brand:  Invitrogen™ C33370

Product Code. 10556393

  • 353.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Click-iT Tetramethylrhodamine (TAMRA) Glycoprotein Detection Kit provides the second part of the simple and robust two-step technique to identify and characterize glycoproteins by one- or two-dimensional gel electrophoresis. In step two, after the incorporation of the azide handle into protein glycan structures with either a Click-iT metabolic labeling reagent or the Click-iT Enzymatic Labeling system, the azide-modified glycoproteins are detected via the chemoselective ligation or click reaction between an azide and an alkyne. Gels with TAMRA labeled glycoproteins can be subsequently stained with the Multiplexed Proteomics™ technologies, SYPRO™ Ruby total protein stain and PRO-Q™ Emerald glycoprotein stain for the differential analysis of glycoprotein subclasses, total proteins and total glycoproteins in the same gel. Click-iT modified glycoproteins are also compatible with downstream LC-MS/MS and MALDI MS analysis for identification.

  • Less hazardous

Cell Analysis, Click Chemistries, Glycoprotein Analysis with Click Chemistry, Labeling Chemistry, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Less hazardous
TAMRA Protein Analysis Detection Kit
Fluorescence
TAMRA™ Isomers, TMR (Tetramethylrhodamine)
Proteins (Glycoproteins)
1 kit
Store in freezer -5°C to -30°C.
Room Temperature
Click-iT™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.