missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Coomassie Fluor™ Orange Protein Gel Stain (Ready-To-Use Solution)

Staining is complete in less than an hour with no risk of overstaining the gel

Brand:  Invitrogen™ C33250

Product Code. 10634613

  • 226.00 € / 1 L
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Molecular Probes proprietary Coomassie Fluor Orange protein gel stain provides fast, simple, sensitive staining of proteins separated by SDS-PAGE. After electrophoresis, the gel is stained, rinsed and photographed. No separate fixation or destaining steps.

Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature and protect from light.
Fluorescence
Protein Gel Stain
1 L
Protein
In-Gel Detection
Coomassie Fluor Orange
Coomassie Fluor™
Room Temperature
300, 470/570 nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.