missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ D-Luciferin, Sodium Salt, 25 mg

Product Code. 11594966
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 11594966

Brand: Invitrogen™ L2912

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Part of the luciferin/luciferase light-producing reaction

The ATP-dependent oxidation of luciferin by luciferase produces light, and this system is employed as a reporter in plants, bacteria, and mammalian cells. Because chemiluminescent techniques are virtually background-free, this reporter gene system is ideal for detecting low-level gene expression.

  • Exhibits physical properties identical to those of natural luciferin
  • Chemiluminescent after oxidation by luciferase
  • Lluciferin sodium salt is readily soluble in aqueous buffers

Find More Fluorogenic and Photoactivatable Probes
A number of fluorogenic substrates, photoactivatable probes, and caged probes are available.

Cell Analysis, Enzyme and Protein Activity Assays, Luciferase Assays, Proteins, Expression, Isolation and Analysis, Reporter Assays

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Type D-Luciferin
Quantity 25 mg
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Chemiluminescence
Shipping Condition Room Temperature
Substrate Properties Chemical Substrate
Substrate Type Luciferase Substrate
Substrate Luciferin

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.