missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DAF-FM Diacetate (4-Amino-5-Methylamino-2',7'-Difluorofluorescein Diacetate)

For detection and quantification of low concentrations of nitric oxide (NO)

Brand:  Invitrogen™ D23844

504.64 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11520276

This item is not returnable. View return policy

Description

Description

It is essentially nonfluorescent until it reacts with NO to form a fluorescent benzotriazole. DAF-FM fluorescence can be detected by any instrument that can detect fluorescein, including flow cytometers, microscopes, fluorescent microplate readers and fluorometers.

  • Ex/Em of DAF-FM: approx. 495/515nm
  • Lyophilized product should be dissolved using DMSO and then added to an aqueous buffer to create a working solution
  • DAF-FM diacetate is cell permeant and passively diffuses across cellular membranes; once inside the cell, it is converted to cell-impermeant for
  • Buffers containing bovine serum albumin (BSA) and phenol red may affect fluorescence and should be used with caution
  • Fluorescence quantum yield of DAF-FM is approx. 0.005, but increases about 160-fold, to ∼0.81, after reacting with NO
  • Assessment of NO production in transaldolase-deficient lymphoblasts by flow cytometry
  • Detection of NO accumulation in embryonic cortical neurons following neurotrophin stimulation
  • in vivo imaging of NO in zebrafish
I

  • ntravital microscopic detection of NO generation associated with angiogenesis in mice
  • Quantitation of ATP-induced NO release in rabbit platelets
  • Spectra of the NO adduct of DAF-FM are independent of pH above pH 5.5
  • NO adduct of DAF-FM is significantly more photostable than that of DAF-2, which means additional time for image capture
  • DAF-FM is a more sensitive reagent for NO than is DAF-2 (NO detection limit for DAF-FM approx. 3nM versus approx. 5nM for DAF-2)

Refer to the Molecular Probes™ Handbook for additional product information.

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Nitric Oxide Research, Nitro-Oxidative Stress

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 x 50 μg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.