Learn More
Invitrogen™ Dextran, Fluorescein, 70,000 MW, Anionic, Lysine Fixable
Rigorous methods for removing as much unconjugated dye as practical, followed by an assay of dextran conjugates by thin-layer chromatography to help ensure the absence of low molecular weight contaminants
Brand: Invitrogen™ D1822
405.84 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of live cells, and to report the hydrodynamic properties of the cytoplasmic matrix. The labeled dextran is commonly introduced into the cells via microinjection. Dextrans are hydrophilic polysaccharides characterized by their moderate-to-high molecular weight, good water solubility, and low toxicity. They also generally exhibit low immunogeniticy. Dextrans are biologically inert due to their uncommon poly-(α-D-1,6-glucose) linkages, which render them resistant to cleavage by most endogenous cellular glycosidases.
- Succinimidyl coupling of dyes to dextran molecule, which, in most cases, results in a neutral or anionic dextran
- Suitable for neuronal tracing (anterograde and retrograde) in live cells
- Cell lineage tracing in live cells
- Examining intercellular communications (e.g., in gap junctions, during wound healing, and during embryonic development); Investigating vascular permeability and blood-brain barrier integrity
- Monitoring acidification (some dextran-dye conjugates are pH-sensitive)
- Studying hydrodynamic properties of the cytoplasmic matrix
- The fluorescein dextrans are intrinsically anionic, whereas most of dextrans labeled with zwitterionic rhodamine B, tetramethylrhodamine, and Texas Red™ dyes are essentially neutral
- To produce more highly anionic dextrans, proprietary procedure adds negatively charged groups to dextran carriers; these products are designated polyanionic dextrans
- Some applications require that dextran tracer be treated with formaldehyde or glutaraldehyde for subsequent analysis: For these applications, lysine-fixable dextran conjugates of fluorophores or biotin
- These dextrans have covalently bound lysine residues that permit dextran tracers to be conjugated to surrounding biomolecules by aldehyde-mediated fixation for subsequent detection by immunohistochemical and ultrastructural techniques
Cell Analysis, Cell Tracing and Tracking, General Cell Tracing, Neuronal Tracing
Order Info
Shipping Condition: Room temperature

Specifications
Dextran | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Invitrogen™ |
25 mg | |
Classic Dyes | |
494/518 nm |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.