missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dextran, Tetramethylrhodamine, 10,000 MW, Lysine Fixable (fluoro-Ruby)

Rigorous methods for removing as much unconjugated dye as practical, followed by an assay of dextran conjugates by thin-layer chromatography to help ensure the absence of low molecular weight contaminants

Brand:  Invitrogen™ D1817

Product Code. 11570216

  • 526.00 € / 25mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of live cells, and to report the hydrodynamic properties of the cytoplasmic matrix. The labeled dextran is commonly introduced into the cells via microinjection. Dextrans are hydrophilic polysaccharides characterized by their moderate-to-high molecular weight, good water solubility, and low toxicity. They also generally exhibit low immunogeniticy. Dextrans are biologically inert due to their uncommon poly-(α-D-1,6-glucose) linkages, which render them resistant to cleavage by most endogenous cellular glycosidases.

  • Succinimidyl coupling of dyes to dextran molecule, which in most cases, results in neutral or anionic dextran
  • Suitable for neuronal tracing (anterograde and retrograde) in live cells
  • Cell lineage tracing in live cells
  • Examining intercellular communications (e.g., in gap junctions, during wound healing, and during embryonic development)
  • Investigating vascular permeability and blood-brain barrier integrity
  • Monitoring acidification (some dextran-dye conjugates are pH-sensitive)
  • Studying the hydrodynamic properties of the cytoplasmic matrix

  • Most of dextrans labeled with tetramethylrhodamine dyes are essentially neutral
  • To produce more highly anionic dextrans, proprietary procedure adds negatively charged groups to dextran carriers; these products are designated polyanionic dextrans
  • Some applications require that dextran tracer be treated with formaldehyde or glutaraldehyde for subsequent analysis: For these applications, lysine-fixable dextran conjugates of fluorophores or biotin
  • These dextrans have covalently bound lysine residues that permit dextran tracers to be conjugated to surrounding biomolecules by aldehyde-mediated fixation for subsequent detection by immunohistochemical and ultrastructural techniques

Cell Analysis, Cell Tracing and Tracking, General Cell Tracing, Neuronal Tracing

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dextran
Store in freezer (-5 to -30°C) and protect from light.
Room Temperature
Invitrogen™
25 mg
Classic Dyes
555/580 nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3123562 Certificate TypeCertificate of Analysis Date16/01/2025 Catalog NumberD1817
Lot Number2795268 Certificate TypeCertificate of Analysis Date13/02/2024 Catalog NumberD1817
Lot Number2544422 Certificate TypeCertificate of Analysis Date27/10/2022 Catalog NumberD1817
Lot Number2441358 Certificate TypeCertificate of Analysis Date29/03/2022 Catalog NumberD1817
Lot Number2403680 Certificate TypeCertificate of Analysis Date14/09/2021 Catalog NumberD1817
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Dextran, Tetramethylrhodamine, 10,000 MW, Lysine Fixable (fluoro-Ruby) > 25mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.