Learn More
Invitrogen™ DiD' solid; DiIC18(5) solid (1,1'-Dioctadecyl-3,3,3',3'-Tetramethylindodicarbocyanine, 4-Chlorobenzenesulfonate Salt)
When applied to cells, dye diffuses laterally within the plasma membrane
Brand: Invitrogen™ D7757
268.28 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
The far-red fluorescent, lipophilic carbocyanine DiD is a longer-wavelength DiI analog. It is supplied as a solid form to facilitate direct application of crystals to membranes and is weakly fluorescent in water but highly fluorescent and quite photostable when incorporated into membranes. It has an extremely high extinction coefficient and short excited-state lifetimes (∽1 nanosecond) in lipid environments.
Cell Analysis, Cell Tracing & Tracking, Neuronal Tracing
Order Info
Shipping Condition: Room Temperature
Specifications
Red | |
Fluorescence Microscope | |
Fluorescence | |
Liphophilic Tracer |
Store at room temperature and protect from light. | |
10 mg | |
Room Temperature | |
Cell Membranes, Lipids |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.