Learn More
Invitrogen™ DiFMUP (6,8-Difluoro-4-Methylumbelliferyl Phosphate)
Continuously assay acid phosphatases at low pH
Brand: Invitrogen™ D6567
Description
DiFMUP is a fluorinated 4-methylumbelliferyl phosphate (MUP) derivative that has a lower pKa than that of MUP, making DiFMUP an excellent substrate for continuously assaying acid phosphatases at low pH. The reaction product of DiFMUP has excitation/emission maxima of approx. 358/450nm.
Biochemical Kinase Assays, Biochemical Toxicology Assays, Cell Analysis, Cellular Imaging, Cellular Toxicology Assays, Drug Discovery and Development, Enzyme and Protein Activity Assays, High-Content Screening (HCS), Kinase Biology, Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation, Target-Based ADME/Tox Assays
Order Info
Shipping Condition: Room temperature
Specifications
Blue | |
Fluorescence | |
5mg | |
Chemical Substrate | |
358⁄455 | |
DiFMU |
Store in freezer (-5 to -30°C). | |
Powder | |
Phosphatase Substrate | |
Phosphatases | |
Microplate Reader | |
Room Temperature |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.