missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dihydrorhodamine 123

Uncharged and nonfluorescent reactive oxygen species indicator

Brand:  Invitrogen™ D23806

329.34 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 10184952

  • 499.00 € / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Dihydrorhodamine 123 can passively diffuse across membranes where it is oxidized to cationic rhodamine 123 which localizes in the mitochondria and exhibits green fluorescence.

Concentration: 5mM

Applications:

Apoptosis, Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Free Radical Detection, Nitro-Oxidative Stress, Reactive Oxygen Species

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.