missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Protease Assay Kit, green fluorescence, 100-1000 assays

Continuously measure protease kinetics

Brand:  Invitrogen™ E6638

Product Code. 10553653

  • 642.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The EnzChek Protease Assay Kit, green fluorescence, is a fast, simple and direct fluorescence-based assay for detecting metallo-, serine, acid, and sulfhydryl proteases. The accompanying increase in fluorescence, which can be measured with fluorescence microplate reader, is proportional to protease activity. This EnzChek Protease Assay Kit contains a casein derivative that has been extensively labeled with the pH-insensitive, green-fluorescent BODIPY FL dye, which results in a quenching of the fluorescent dye. Protease-catalyzed hydrolysis releases the highly-fluorescent BODIPY FL dye-labeled peptides, allowing for quantitative detection of protease activity in solution. The green-fluorescent BODIPY FL dye has excitation and emission spectra similar to those of fluorescein.

  • Generalized mechanism allows for measurement of a variety of proteases
  • Can be used to continuously measure protease kinetics
  • Excitation/emission of 505/513nm allows use of standard fluorescein (FITC) filter settings

Cell Analysis, Cell Culture, Cell Viability, Proliferation and Function, Differentiation and Development, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis, Stem Cell Analysis, Stem Cell Differentiation, Stem Cell Protein Analysis, Stem Cell Research

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer -5°C to -30°C and protect from light.
Protease Assay
Protease Assay
1 kit
Room Temperature
Protease Substrate
Fluorescence Intensity
Fluorescence Microplate Reader
EnzChek™
Differentiation
Protein-Based Substrate
Proteases
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.