missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Epidermal Growth Factor, Biotinylated, complexed to Alexa Fluor™ 488 Streptavidin (Alexa Fluor™ 488 EGF complex)

Product Code. 11598946
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 11598946

Brand: Invitrogen™ E13345

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The biotinylated conjugates utilize biotin-XX, which contains a long spacer arm designed to enhance the probe affinity for the EGF receptor

Fluorescently labeled EGF has enabled scientists to investigate receptor-membrane interactions, study receptor distribution, calculate rate constants for the interaction of EGF with its receptor, and more.

  • Cconjugates incorporating fluorescein (FITC), Oregon Green™ 514, and tetramethylrhodamine were constructed with dye directly attached to EGF
  • EGF molecule: 53 amino acids, MW=6,045 Da
  • Label (Ex/Em): Biotin-XX complexed with Alexa Fluor 488 streptavidin (approx. 495/519nm)
  • Number of fluorophore molecules on each EGF molecule: approximately 2 to 3
  • Fluorescence is typically monitored using a flow cytometer, fluorescence microscope, or fluorimeter

Bioproduction, Cell Culture, Mammalian Cell Culture, Primary Cell Culture, T Cell Culture

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mammalian Cell
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Product Type Growth Factor
Dye Type Alexa Fluor Dyes
Form Solid
Product Line Alexa Fluor
Quantity 100 μg
Shipping Condition Wet Ice
Protein Family EGF

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.