missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ethidium Homodimer-2 (EthD-2) - 1 mM Solution in DMSO

Cell-impermeant viability indicator

Brand:  Invitrogen™ E3599

628.90 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 10003332

  • 678.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ethdium homodimer-2 (EthD-2) is a high-affinity nucleic acid stain that is weakly fluorescent until bound to DNA and emits red fluorescence (excitation/emission maxima ∽535/624).

Cell Analysis, Cell Structure, Cell Tracing & Tracking, Cell Viability & Cytotoxicity, Cell Viability, Proliferation & Function, Microbial Tracking, Nucleus, Nucleoli & Nuclear Envelope, Organelle Tracing

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Orange
535 nm
Cell staining assays
200 μL
Ethidium Homodimer-2 (EthD-2) - 1 mM Solution in DMSO
Room Temperature
Viability Indicator
Store in freezer at -5°C to -30°C and protect from light.
Cell-Impermeant
Fluorescence Microscope
Fluorescence
624 nm
Fluorescent Dye
Nucleic Acids, Nucleus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.