missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EZQ™ Protein Quantitation Kit

Fluorescence-based protein assay facilitates fast quantitation of protein samples prepared for gel electrophoresis

Brand:  Invitrogen™ R33200

Product Code. 10780184

  • 810.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Assay can be performed in presence of detergents, urea and reducing agents—simply spot 1μL of protein sample onto prepared paper, stain with proprietary fluorescent dye, and measure fluorescence
  • Samples can be quantitated by comparison with standard curve
  • Specially-designed 96-well microplate for easy quantitation of samples on microplate reader or laser scanner provided for added versatility

Protein Quantitation, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature and protect from light.
Fluorescence
Protein Quantitation Assay
Protein Quantitation Assay
2000 Assays
Store at room temperature. Protect from light.
Fluorescent Protein Assay
EZQ™
2000 Assays
Microplate Reader
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.