Learn More
Invitrogen™ FluoCells™ Prepared Slide #1 (BPAE cells with MitoTracker™ Red CMXRos, Alexa Fluor™ 488 Phalloidin, and DAPI)
Contains fixed, multi-labeled, cell preparations for observation by fluorescence microscopy validated for immunofluorescence
Brand: Invitrogen™ F36924
201.40 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Description
FluoCells Prepared Slide #1 contains bovine pulmonary artery endothelial cells (BPAEC). MitoTracker Red CMXRos was used to stain the mitochondria in the live cells, with accumulation dependent upon membrane potential. Following fixation and permeabilization, F-actin was stained with Alexa Fluor 488 phalloidin, and the nuclei were counterstained with the blue-fluorescent DNA stain DAPI.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF)
Order Info
Shipping Condition: Room temperature

Specifications
Prepared Microscope Slide | |
Immunofluorescence | |
1 Ea. |
Alexa Fluor FluoCells MitoTracker | |
Store at room temperature and protect from light. | |
Room Temperature |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.