missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescein Di-β-D-Glucopyranoside (FDGlu)

Cell permeant

Brand:  Invitrogen™ F2881

261.44 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 11514816

  • 348.00 € / 5mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Fluorescein di-β-D-glucopyranoside is one of the most sensitive substrates for glucosidases. The nonfluorescent substrate is sequentially hydrolyzed by β-glucosidase, first to fluorescein monoglucoside and then to highly fluorescent fluorescein. Enzyme-mediated hydrolysis of fluorescein di-β-D-glucopyranoside can be followed by the increase in either absorbance or fluorescence. Sensitive substrate for glucosidases.

Cell Analysis, Enzyme and Protein Activity Assays, Flow Cytometry, Glycosidase, Neuraminidase, and Related Enzyme Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell-Permeant
Fluorescence
Beta-Glucosidase Substrate
490⁄514
Store in freezer (-5 to -30°C).
5 mg
Chemical Substrate
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2984202 Certificate TypeCertificate of Analysis Date31/07/2024 Catalog NumberF2881
Lot Number2984202 Certificate TypeCertificate of Analysis Date29/07/2024 Catalog NumberF2881
Lot Number2541670 Certificate TypeCertificate of Analysis Date18/08/2023 Catalog NumberF2881
Lot Number2403746 Certificate TypeCertificate of Analysis Date23/10/2021 Catalog NumberF2881
Lot Number2342909 Certificate TypeCertificate of Analysis Date08/06/2021 Catalog NumberF2881
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Fluorescein Di-β-D-Glucopyranoside (FDGlu) > 5mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.