missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescein Diphosphate, Tetraammonium Salt (FDP)

Colorless and non-fluorescent

Brand:  Invitrogen™ F2999

Product Code. 11534816

  • 726.00 € / 5mg
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

FDP is a colorless and nonfluorescent substrate for alkaline phosphatases. Sequential alkaline phosphatase mediated hydrolysis of its two phosphate substituents yields weakly fluorescent fluorescein monophosphate followed by strongly fluorescent fluorescein (excitation/emission approx. 490/514nm).

Alkaline Phosphatase Assays, Cell Analysis, Drug Discovery and Development, Enzyme and Protein Activity Assays, Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Reporter Assays, Target and Lead Identification and Validation

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer (-5 to -30°C).
Powder
Phosphatase Substrate
Alkaline Phosphatase, Tyrosine Phosphatases
Fluorometer, Microplate Reader
Room Temperature
Fluorescence
5 mg
Chemical Substrate
490⁄514
FITC (Fluorescein)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.