missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Escherichia coli (K-12 strain) BioParticles™, fluorescein conjugate
The fluorescence of the dye conjugates is uniformly intense over the pH range from 3 to 10.
Brand: Invitrogen™ E2861
286.52 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
- Conjugates covalently labeled with variety of different fluorophores (special care has been taken to remove free dye after conjugation)
- Provided as lyophilized powders
- Approximately 3 x 108 E. coli or S. aureus particles per mg solid and approximately 2 x 107 zymosan particles per mg solid
- Conjugates can be reconstituted in buffer of choice for use in phagocytosis assays
- Fluorescence of conjugates that are bound to surface of cell (but not internalized) can be quenched by ethidium bromide, trypan blue, or other quenchers
- Addition to cellular applications, fluorescent conjugates may be effective as flow cytometry calibration references when sorting bacteria and yeast mutants
- Small particles may also be useful references for light scattering studies because their sizes and shapes differ in characteristic ways
Order Info
Shipping Condition: Room Temperature
Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
Fluorescence Microscope | |
FITC (Fluorescein) | |
3 to 10 | |
10 mg | |
E. coli |
Fluorescence | |
E.coli Conjugate | |
Solid | |
BioParticles™ | |
Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction