Learn More
Invitrogen™ Image-IT™ LIVE Mitochondrial Transition Pore Assay Kit, for microscopy
For microscopy
Brand: Invitrogen™ I35103
1130.50 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
The Image-iT LIVE Mitochondrial Transition Pore Assay Kit provides a proven method [REF PN36389 ,PN32679] of measuring mitochondrial permeability transition pore opening. The kit includes a dye and a quencher that are easily loaded into cells. In healthy cells, the dye, but not the quencher, enters mitochondria. The mitochondria remain brightly fluorescent until mitochondrial pore activation permits quenching of the fluorescence.
Apoptosis, Cell Analysis, Cell Structure, Cell Viability, Proliferation & Function, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Mitochondria, Mitochondria Function, Mitochondrial Function
Order Info
Shipping Condition: Room Temperature
Specifications
1 Kit | |
Mitochondrial Transition Pore Assay Kit | |
Orange, Blue, Green | |
Mitochondria | |
Fluorescence Microscope |
Kit | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Image-iT™ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.