Learn More
Invitrogen™ HCS LipidTOX™ Green Phospholipidosis Detection Reagent (1000X), for cellular imaging
Easily detected by fluorescence microscopy
Marca: Invitrogen™ H34350
325.50 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Descripción
The HCS LipidTOX Green phospholipidosis detection reagent was developed to characterize the potentially toxic side effects of compounds on lipid metabolism in mammalian cell lines. This reagent is easily detected by fluorescence microscopy or an HCS reader equipped with filters that can detect excitation/emission maxima approx. 495/525nm. LipidTOX phospholipidosis detection reagents do not affect the normal growth of cells, and the staining observed after incubation in live cells is maintained after formaldehyde fixation. This probe is compatible with the HCS LipidTOX neutral lipid stains.
Cell Analysis, Cellular Imaging, High-Content Screening (HCS)
Información de órdenes
Shipping Condition: Room temperature

Especificaciones
Store in freezer -5°C to -30°C and protect from light. | |
High Content Analysis Instrument | |
Other Label(s) or Dye(s) | |
LipidTOX™ | |
Room Temperature |
Fluorescence | |
Phospholipidosis Detection Reagent | |
96-well plate | |
Each |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only. Not for use in diagnostic procedures.