missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NBD-PE (N-(7-Nitrobenz-2-Oxa-1,3-Diazol-4-yl)-1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt)

The phospholipid, NBD-PE is labeled on the head group with the environment sensitive fluorophore, NBD.

Brand:  Invitrogen™ N360

248.50 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 11509136

  • 355.00 € / 10mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The phospholipid, NBD-PE is labeled on the head group with the environment sensitive fluorophore, NBD (excitation/emission maxima ∼463/536 nm).

Prepare stock solutions of phosphoethanolamines in ethanol to attain a stock concentration of 1 to 2 mg/mL. Sonication may be necessary to aid dispersion.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospholipids
956.25
Invitrogen
10 mg
Solid
463/536 nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3123527 Certificate TypeCertificate of Analysis Date05/01/2025 Catalog NumberN360
Lot Number2984229 Certificate TypeCertificate of Analysis Date22/07/2024 Catalog NumberN360
Lot Number2833429 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberN360
Lot Number2500561 Certificate TypeCertificate of Analysis Date09/02/2023 Catalog NumberN360
Lot Number2441495 Certificate TypeCertificate of Analysis Date21/02/2022 Catalog NumberN360
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ NBD-PE (N-(7-Nitrobenz-2-Oxa-1,3-Diazol-4-yl)-1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt) > 10 mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.