Learn More
Invitrogen™ Nitro blue Tetrazolium Chloride (NBT)
Use to assess cell viability as function of redox potential
Brand: Invitrogen™ N6495
238.64 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
298.30 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
- Actively respiring cells convert water-soluble Nitro blue tetrazolium chloride to a deep blue-colored precipitate
Cell Analysis, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cellular Imaging, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunohistochemistry (IHC), Nitro-Oxidative Stress, Proteins, Expression, Isolation and Analysis, Reactive Oxygen Species, Western Blotting
Order Info
Shipping Condition: Room Temperature

Specifications
Store at room temperature and protect from light. | |
1 g | |
Chemical Substrate |
Colorimetric | |
Oxidase Substrate | |
Room Temperature |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.