missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NucBlue™ Live ReadyProbes™ Reagent (Hoechst 33342)

Hoechst stain formulated in room temperature-stable solution that is provided in dropper bottle

Brand:  Invitrogen™ R37605

Product Code. 12303553

  • 177.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Hoechst 33342 is perhaps the most ubiquitous nuclear fluorescence stain for live and fixed cells and tissue sections. The cell permeable stain binds preferentially to AT regions of DNA. It is excited by UV light and emits blue fluorescence at 460nm when bound to DNA.

  • Ready-to-use liquid formulation of high-purity Hoechst 33342
  • Typically Hoechst 33342 is supplied as solid or highly concentrated solution that requires weighing out and diluting several thousand-fold before use, and recommended storage is generally in freezer or refrigerator
  • This version is ready to use and can be stored next to a microscope or workstation. A liquid formulation in ultra-convenient dropper bottle allows you to see
  • Spectral properties of Hoechst 33342 (2'-[4-ethoxyphenyl]-5-[4-methyl-1-piperazinyl]-2,5'-bi-1H-benzimidazole), including a large Stokes shift, make it ideal for use with green (Alexa Fluor 488, FITC, GFP) and red (Alexa Fluor 594, Texas Red, rhodamine, mCherry, mKate-2) fluorophores in multicolor experiments
  • Because of its high affinity to DNA, Hoechst 33342 is also frequently used in cell counting, cell cycle, and cell replication studies to distinguish condensed nuclei in apoptotic cells, for cell-cycle studies, as a nuclear segmentation tool in high content imaging analysis, and to sort cells based on their DNA content
  • May be added directly to cells in full media, or buffer solutions
  • In most cases 2 drops/mL and incubation of 15 to 30 minutes will give bright nuclear staining; however, optimization may be needed for some cell types, conditions, and applications
  • In most cases, staining intensity increases with time if cells are not washed prior to imaging.
  • Excited by UV light at 360nm when bound to DNA, with an emission maximum at 460nm
  • Detected through blue/cyan filter

Apoptosis, Cell Analysis, Cell Cycle, Cell Proliferation, Cell Structure, Cell Tracing and Tracking, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cellular Imaging, DNA Fragmentation, Flow Cytometry, Flow Cytometry Staining by Cell Structure, Immunofluorescence (IF), Immunofluorescence Counterstaining, Mounting and Fade Prevention, Immunofluorescence Staining and Detection, Nucleus, Nucleoli and Nuclear Envelope, Organelle Tracing

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blue
360⁄460
Fluorescence Microscope, Flow Cytometer
Fluorescence
Liquid
Room Temperature
Nucleic Acid Stain
6 × 2.5 mL dropper bottles

Store at ≤ 25°C

Hoechst 33342
1 kit
Visible
Molecular Probes™
Fluorescent Dye
Nucleus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.